Order Entry
Export
Orders LinkContactUsLinkComponent
 

Tetramethylsilane, Sigma-Aldrich®

Flammable
Supplier:   Merck
Tetramethylsilane, Sigma-Aldrich®
Supplier:  Merck
Certificates
Tetramethylsilane, Sigma-Aldrich®

2 Options Available Below

Product Details & Documents
Synonyms:  TMS
  • CAS Number:75-76-3
  • Formula:C₄H₁₂Si
  • MDL Number:MFCD00008274
  • Molecular Weight:88.22 g/mol
  • Boiling Pt:26.6 °C (1013 hPa)
  • Melting Pt:–99 °C
  • Density:0.642 g/cm³ (20 °C)
  • Flash Pt:–54 °C
  • Storage Temperature:Refrigerator
  • EINECS:200-899-1
  • ADR:3,I
  •  :M-Clarity™ quality level = MQ200
  • UN:2749
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 78,00 €
    78,00 €
    1 * 25 g
       78,00 €

  • Glass bottle for solids
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 150,00 €
    150,00 €
    1 * 100 g
       150,00 €