Order Entry
Export
Orders LinkContactUsLinkComponent
 

Tetramethylsilane 99%

Tetramethylsilane 99%
 
Tetramethylsilane 99%
Tetramethylsilane 99%

 

 
 :  TMS
  •  :75-76-3
  •  :C₄H₁₂Si
  •  :MFCD00008274
  •  :88.22 g/mol
  •  :26.6 °C (1013 hPa)
  •  :–99 °C
  •  :0.64 g/cm³
  •  :–27 °C
  •  :Refrigerator
  •  :200-899-1
  •  :2749
  •  :3,I

 

 

 

  • Appearance:
    Clear colorless Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.3575 to 1.3595 (20°C, 589 nm)
  • GC:
    ≥98.5 %
Product Family Options

 

  • Pack type
    Pk
     
     
  • Glass bottle
    100 mL
       
       
    1 * 100 mL
    1 * 100 mL - 77,30 €
    77,30 €
    1 * 100 mL
       77,30 €

  • Glass bottle
    500 mL
       
       
    1 * 500 mL
    1 * 500 mL - 312,00 €
    312,00 €
    1 * 500 mL
       312,00 €

  • Glass bottle
    2,5 L
       
       
    1 * 2,5 L
    1 * 2,5 L - 1 230,00 €
    1 230,00 €
    1 * 2,5 L
       1 230,00 €