Order Entry
Export
Orders LinkContactUsLinkComponent
Methyldiphenylsilane ≥97%

Methyldiphenylsilane ≥97%

Methyldiphenylsilane ≥97%
Certificates
Methyldiphenylsilane ≥97%
Methyldiphenylsilane ≥97%

2 Options Available Below

Product Details & Documents

Purity: 97% CAS No.: 776-76-1 Molecular Formula: C13H14Si Formula Weight: 198.34 Flash Point: 120°(248°F) Refractive Index: 1.5720 Odour: Odourless Boiling Point: 266-267° Beilstein Reference: 1867183 MDL No.: MFCD00008255

Synonyms:  Diphenylmethylsilane
  • CAS Number:776-76-1
  • Formula:C₁₃H₁₄Si
  • MDL Number:MFCD00008255
  • Molecular Weight:198.34 g/mol
  • Boiling Pt:266…267 °C
  • Density:1.001
  • Flash Pt:120 °C (248 °F)
  • Storage Temperature:Ambient
  • EINECS:212-281-9
Product Family Options

Product Information