

(3-Aminopropyl)triethoxysilane ≥98%, Sigma-Aldrich®
:919-30-2
:
:H₂NCH₂CH₂CH₂Si(OCH₂CH₃)₃
:MFCD00008207
:221.37 g/mol
:217 °C (1013 hPa)
:220...222 °C
:0.95 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
:93 °C (closed cup)
:Room Temperature
:213-048-4
:GHS05
:GHS07
- Appearance (Colour):Colorless to Very Faint Yellow
- Appearance (Form):Liquid
- Appearance (Turbidity):Clear
- Proton NMR Spectrum:Conforms to Structure
- Titration by HClO₄:≥98%
- Recommended Retest Period:2 years