

(3-Aminopropyl)triethoxysilane, Sigma-Aldrich®
Catalog # SIAL706493-20ML
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:20 mL
- Seq:2305
Specifications
About this item
:
:221.37 g/mol
Boiling Pt:217 °C (1013 hPa)
Melting Pt:< –70 °C
Density:0.95 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
Flash Pt:104 °C
EINECS:213-048-4
:GHS05
:GHS07
Storage Temperature:Ambient
CAS Number:919-30-2
Formula:H₂NCH₂CH₂CH₂Si(OCH₂CH₃)₃
MDL Number:MFCD00008207