

(3-Aminopropyl)triethoxysilane
Supplier: Roth Carl
Certificates
About this item
CAS Number:919-30-2
:
Formula:H₂NCH₂CH₂CH₂Si(OCH₂CH₃)₃
MDL Number:MFCD00008207
:221.37 g/mol
Boiling Pt:217 °C (1013 hPa)
Melting Pt:< –70 °C
Density:0,95 g/cm³ (20 °C)
Flash Pt:104 °C
Storage Temperature:Ambient
EINECS:213-048-4
:GHS05
:GHS07