Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
(3-Aminopropyl)triethoxysilane
(3-Aminopropyl)triethoxysilane 98%
(3-Aminopropyl)triethoxysilane 98%
:
Thermo Fisher Scientific
(3-Aminopropyl)triethoxysilane 98%
:
Thermo Fisher Scientific
:
919-30-2
:
:
H₂NCH₂CH₂CH₂Si(OCH₂CH₃)₃
:
MFCD00008207
:
221.37 g/mol
:
217 °C
:
< –70 °C
:
0.948
:
104 °C (219 °F)
:
Ambient
:
213-048-4
:
8,II
:
GHS05
:
GHS07
msds_ALFAA10668
Thermo Scientific Acroseal 25 Years