Order Entry
Export
ContactUsLinkComponent
Silicon dioxide, Sigma-Aldrich®
Silicon dioxide, Sigma-Aldrich®
Catalog # SIAL85356-100G
Supplier:  Merck
CAS Number:  
Silicon dioxide, Sigma-Aldrich®
Catalog # SIAL85356-100G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :60.08 g/mol
Merck Index:14,08493
Boiling Pt:2230 °C (1013 hPa)
Melting Pt:1719 °C
Density:2.2 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-545-4
CAS Number:7631-86-9
Formula:SiO₂
MDL Number:MFCD00011232