Order Entry
Export
Orders LinkContactUsLinkComponent
Lead(IV) oxide 97+%, extra pure

Lead(IV) oxide 97+%, extra pure

Irritant
Health
Environment
Oxidizing
Lead(IV) oxide 97+%, extra pure
Certificates
Lead(IV) oxide 97+%, extra pure

4 Options Available Below

Product Details & Documents
Synonyms:  Lead dioxide
  • CAS Number:1309-60-0
  • Formula:O₂Pb
  • MDL Number:MFCD00011165
  • Molecular Weight:239.2 g/mol
  • Merck Index:12,05424
  • Melting Pt:290 °C (dec.)
  • Density:9,4 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:215-174-5
  • UN:1872
  • ADR:5.1,III

Documents

Specifications for Product Family

Specification Test Results

  • Appearance:
    Brown to black Powder
  • Titration with Na2S2O3:
    ≥97.0 %
  • Chloride (Cl):
    ≤0.02 %
  • Sulfate (SO4):
    ≤0.005 %
  • Copper (Cu):
    ≤0.01 %
  • Iron (Fe):
    ≤0.01 %
  • Zinc (Zn):
    ≤0.005 %
Product Family Options

Product Information