Lead(II) oxide
Glass bottle for solids, 250 g
Catalog # ROTH4456.1
Supplier: Roth Carl
Jump to:
Product Details & Documents
Synonyms: Lead monoxide
- CAS Number:1317-36-8
- Formula:PbO
- MDL Number:MFCD00011164
- Molecular Weight:223.2 g/mol
- Merck Index:13,05431
- Boiling Pt:1470 °C (1013 hPa)
- Melting Pt:888 °C
- Density:9.5 g/cm³ (20 °C)
- EINECS:215-267-0
- UN:2291
- ADR:6.1,III
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:250 g



