Order Entry
Export
Orders LinkContactUsLinkComponent
Lead(II) iodide ≥98.5%

Lead(II) iodide ≥98.5%

Health
Irritant
Environment
Lead(II) iodide ≥98.5%
Certificates
Lead(II) iodide ≥98.5%
Lead(II) iodide ≥98.5%

2 Options Available Below

Product Details & Documents

Purity: 98.5% CAS No.: 10101-63-0 Molecular Formula: I2Pb Formula Weight: 461.00 Physical Form: Powder Density: 6.16 Appearance: Yellow Odour: Odourless Melting Point: 402° Boiling Point: 954° Solubility: Partially soluble in water. Freely soluble in sodium thiosulfate solution. Soluble in concentrated solutions of alkali iodides. Insoluble in alcohol and cold HCl Merck Reference: 14,5411 UN No.: 2291 MDL No.: MFCD00011163

Synonyms:  Lead diiodide
  • CAS Number:10101-63-0
  • Formula:I₂Pb
  • MDL Number:MFCD00011163
  • Molecular Weight:461.01 g/mol
  • Merck Index:14,5411
  • Boiling Pt:954 °C
  • Melting Pt:402 °C
  • Density:6.16
  • Storage Temperature:Ambient
  • EINECS:233-256-9
  • UN:2291
  • ADR:6.1,III
Product Family Options

Product Information