Lead(II) iodide 99%
Supplier: Thermo Fisher Scientific
About this item
Synonyms: Lead diiodide
- CAS Number:10101-63-0
- Formula:I₂Pb
- MDL Number:MFCD00011163
- Molecular Weight:461.01 g/mol
- Merck Index:12,05430
- Boiling Pt:954 °C (1013 hPa)
- Melting Pt:402 °C
- Density:6.16 g/cm³
- EINECS:233-256-9
- UN:2291
- ADR:6.1,III
2 Options Available Below
Product Details & Documents
Synonyms: Lead diiodide
- CAS Number:10101-63-0
- Formula:I₂Pb
- MDL Number:MFCD00011163
- Molecular Weight:461.01 g/mol
- Merck Index:12,05430
- Boiling Pt:954 °C (1013 hPa)
- Melting Pt:402 °C
- Density:6.16 g/cm³
- EINECS:233-256-9
- UN:2291
- ADR:6.1,III
Specifications for Product Family
Specification Test Results
- Appearance:Yellow to orange Powder
- Titration Argentometric:≥98.5 %
- Loss on drying:≤0.1 %
- Chloride (Cl):≤100 ppm



