Order Entry
Export
Orders LinkContactUsLinkComponent
Lead(IV) fluoride ≥99%

Lead(IV) fluoride ≥99%

Plastic bottle for solids, 2 g
Catalog # 13070.04
Health
Irritant
Environment
Lead(IV) fluoride ≥99%
Catalog # 13070.04
CAS Number:  7783-59-7
Lead(IV) fluoride ≥99%
Lead(IV) fluoride ≥99%

Plastic bottle for solids, 2 g

CAS Number:  7783-59-7
Price
129,00 €
Log in for availability
1 * 2 g
Product Details & Documents

Purity: 99% CAS No.: 7783-59-7 Molecular Formula: PbF4 Formula Weight: 283.18 Physical Form: Crystalline Sensitivity: Moisture Sensitive UN No.: 2291 MDL No.: MFCD00016278

Synonyms:  Lead tetrafluoride
  • CAS Number:7783-59-7
  • Formula:F₄Pb
  • MDL Number:MFCD00016278
  • Molecular Weight:283.19 g/mol
  • Merck Index:12,05448
  • Melting Pt:600 °C
  • Density:6.7 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:232-012-9
  • UN:2291
  • ADR:6.1,III
  •  :Density: 6.7 g/cm³ (20 °C)
Specifications

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    2 g

Specifications