Lead(II) oxide ≥99.99% (metals basis)
Supplier: Thermo Fisher Scientific
Jump to:
About this item
Purity: 99.99% CAS No.: 1317-36-8 Molecular Formula: OPb Formula Weight: 223.19 Physical Form: Powder Density: 9.53 Appearance: Yellow Odour: Odourless Melting Point: 886° Merck Reference: 14,5413 UN No.: 3077 MDL No.: MFCD00011164
Synonyms: Lead monoxide
- CAS Number:1317-36-8
- Formula:PbO
- MDL Number:MFCD00011164
- Molecular Weight:223.2 g/mol
- Merck Index:14,5413
- Boiling Pt:1470 °C (1013 hPa)
- Melting Pt:886 °C
- Density:9.53
- Storage Temperature:Ambient
- EINECS:215-267-0
- UN:3077
- ADR:9,III
3 Options Available Below
Product Details & Documents
Purity: 99.99% CAS No.: 1317-36-8 Molecular Formula: OPb Formula Weight: 223.19 Physical Form: Powder Density: 9.53 Appearance: Yellow Odour: Odourless Melting Point: 886° Merck Reference: 14,5413 UN No.: 3077 MDL No.: MFCD00011164
Synonyms: Lead monoxide
- CAS Number:1317-36-8
- Formula:PbO
- MDL Number:MFCD00011164
- Molecular Weight:223.2 g/mol
- Merck Index:14,5413
- Boiling Pt:1470 °C (1013 hPa)
- Melting Pt:886 °C
- Density:9.53
- Storage Temperature:Ambient
- EINECS:215-267-0
- UN:3077
- ADR:9,III



