Lead(II) chloride, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:7758-95-4
:
Formula:Cl₂Pb
MDL Number:MFCD00011157
:278.11 g/mol
Merck Index:13,05422
Boiling Pt:950 °C (1013 hPa)
Melting Pt:496 °C
Density:5.85 g/cm³ (20 °C)
Storage Temperature:Ambient
UN:2291
ADR:6.1,III
:M-Clarity™ quality level = MQ200
EINECS:231-845-5
:GHS08
:GHS07
:GHS09