Order Entry
Export
Orders LinkContactUsLinkComponent
 

Lead(II) bromide ≥98%

Health
Irritant
Environment
Lead(II) bromide ≥98%
Certificates
Lead(II) bromide ≥98%
Lead(II) bromide ≥98%

2 Options Available Below

Product Details & Documents

Purity: 98+% CAS No.: 10031-22-8 Molecular Formula: Br2Pb Formula Weight: 367.01 Odour: Odourless Melting Point: 373° Boiling Point: 916° Merck Reference: 14,5403 UN No.: 2291 MDL No.: MFCD00011156

Synonyms:  Lead dibromide, Lead(II) bromide
  • CAS Number:10031-22-8
  • Formula:Br₂Pb
  • MDL Number:MFCD00011156
  • Molecular Weight:367.01 g/mol
  • Merck Index:14,5403
  • Boiling Pt:916 °C
  • Melting Pt:373 °C
  • Density:6.66
  • Storage Temperature:Ambient
  • EINECS:233-084-4
  • UN:2291
  • ADR:6.1,III
Product Family Options

Product Information