Order Entry
Export
Orders LinkContactUsLinkComponent
 

Lead(II) bromide 98+%, extra pure

Irritant
Health
Environment
Lead(II) bromide 98+%, extra pure
Certificates
Lead(II) bromide 98+%, extra pure

2 Options Available Below

Product Details & Documents
Synonyms:  Lead dibromide, Lead(II) bromide
  • CAS Number:10031-22-8
  • Formula:Br₂Pb
  • MDL Number:MFCD00011156
  • Molecular Weight:367.01 g/mol
  • Merck Index:12,05418
  • Boiling Pt:916 °C (1013 hPa)
  • Melting Pt:373 °C
  • Density:6.66 g/cm³
  • EINECS:233-084-4
  • UN:2291
  • ADR:6.1,III

Documents

Specifications for Product Family

Specification Test Results

  • Appearance:
    White to off-white Crystalline powder
  • Titration Argentometric:
    ≥98.0 %
  • Loss on drying:
    ≤0.1 %
  • Chloride (Cl):
    ≤100 ppm
  • Insoluble matter:
    ≤0.005 % (in acid)
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 32,70 €
    32,70 €
    1 * 100 g
       32,70 €

  • Glass bottle for solids
    500 g
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 107,00 €
    107,00 €
    1 * 500 g
       107,00 €