
Aluminium ammonium sulphate dodecahydrate 99%, pure
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7784-26-1
:MFCD00149958
Formula:AlNH₄(SO₄)₂·12H₂O
:453.33 g/mol
Merck Index:13,00325
Boiling Pt:200 °C (760 mmHg)
Melting Pt:93.5 °C
Density:1.64 g/cm³
Storage Temperature:Ambient
EINECS:232-055-3
:GHS07
Specification Test Results
- Appearance:White Crystalline powder or crystals
- Titration Complexometric:98.5 to 102.5 %
- Loss on drying:45 % to 48 % (2 g, 300°C)