Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Aluminium oxide
Aluminium oxide substrate 10x10x1 mm, polished one side C plane
Aluminium oxide substrate 10x10x1 mm, polished one side C plane
:
Thermo Fisher Scientific
Aluminium oxide substrate 10x10x1 mm, polished one side C plane
:
Thermo Fisher Scientific
:
1344-28-1
:
11092-32-3
:
Al₂O₃
:
MFCD00003424
:
101.96 g/mol
:
13,00355
:
2980 °C (1013 hPa)
:
2045 °C
:
3.94 g/cm³ (20 °C)
:
Ambient
:
215-691-6
:
01-2119529248-35
msds_ALFA040281
Thermo Scientific Acroseal 25 Years