Aluminium oxide polishing compound
Catalog # 39807.LA
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:1
- Seq:0001
Specifications
About this item
:
:101.96 g/mol
Merck Index:14,356
Boiling Pt:2980 °C
Melting Pt:2045 °C
Density:3.94 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:215-691-6
REACH:01-2119529248-35
CAS Number:1344-28-1
:11092-32-3
Formula:Al₂O₃
MDL Number:MFCD00003424