Order Entry
Export
Orders LinkContactUsLinkComponent
Barium sulphate

Barium sulphate

Glass bottle for solids, 1 kg
Catalog # MOLE21615331-1KG
Supplier:   Molekula
Barium sulphate
Catalog # MOLE21615331-1KG
Supplier:  Molekula
CAS Number:  7727-43-7
Barium sulphate

Glass bottle for solids, 1 kg

CAS Number:  7727-43-7
Price
92,00 €
Log in for availability
1 * 1 kg
Product Details & Documents
  • CAS Number:7727-43-7
  • Formula:BaSO₄
  • MDL Number:MFCD00003455
  • Molecular Weight:233.39 g/mol
  • Merck Index:13,00997
  • Boiling Pt:1600 °C (1013 hPa)
  • Melting Pt:1345 °C
  • Density:4.5 g/cm³ (25 °C)
  • Storage Temperature:Ambient
  • EINECS:231-784-4
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg

Specifications