Order Entry
Export
Orders LinkContactUsLinkComponent
Calcium hydroxide

Calcium hydroxide

Corrosive
Irritant
Supplier:   Roth Carl
Calcium hydroxide
Supplier:  Roth Carl
Certificates
Calcium hydroxide

4 Options Available Below

Product Details & Documents
Synonyms:  Slaked lime, Calcium dihydroxide
  • CAS Number:1305-62-0
  • Formula:Ca(OH)₂
  • MDL Number:MFCD00010901
  • Molecular Weight:74.09 g/mol
  • Merck Index:13,01675
  • Melting Pt:550 °C
  • Density:2.24 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:215-137-3
  • UN:3262
  • ADR:8,III
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    1 kg
    Log in for availability
    Log in for availability
    1 * 1 kg
    1 * 1 kg - 19,80 €
    19,80 €
    1 * 1 kg
       19,80 €

  • Glass bottle for solids
    2,5 kg
    Log in for availability
    Log in for availability
    1 * 2,5 kg
    1 * 2,5 kg - 42,10 €
    42,10 €
    1 * 2,5 kg
       42,10 €

  • 10 kg
    Log in for availability
    Log in for availability
    1 * 10 kg
    1 * 10 kg - 70,50 €
    70,50 €
    1 * 10 kg
       70,50 €

  • 50 kg
    Log in for availability
    Log in for availability
    1 * 50 kg
    1 * 50 kg - 145,00 €
    145,00 €
    1 * 50 kg
       145,00 €