Order Entry
Export
Orders LinkContactUsLinkComponent
Barium oxide, Sigma-Aldrich®

Barium oxide, Sigma-Aldrich®

Toxic
Corrosive
Supplier:   Merck
Barium oxide, Sigma-Aldrich®
Supplier:  Merck
Certificates
Barium oxide, Sigma-Aldrich®

2 Options Available Below

Product Details & Documents
  • CAS Number:1304-28-5
  • Formula:BaO
  • MDL Number:MFCD00003453
  • Molecular Weight:153.33 g/mol
  • Merck Index:12,01015
  • Boiling Pt:∼2000 °C (1013 hPa)
  • Melting Pt:∼1920 °C
  • Density:∼5.72 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:215-127-9
  • ADR:6.1,III
  •  :M-Clarity™ quality level = MQ100
  • UN:1884
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 59,70 €
    59,70 €
    1 * 5 g
       59,70 €

  • Glass bottle for solids
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 214,00 €
    214,00 €
    1 * 25 g
       214,00 €