Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Calcium hydride
Calcium hydride, Sigma-Aldrich®
Calcium hydride, Sigma-Aldrich®
:
Merck
Calcium hydride, Sigma-Aldrich®
:
Merck
:
7789-78-8
:
:
H₂Ca
:
MFCD00010897
:
42.09 g/mol
:
13,01674
:
816 °C
:
1.9 g/cm³ (20 °C)
:
232-189-2
:
4.3,I
:
M-Clarity™ quality level = MQ100
:
1404
:
GHS02
:
Ambient