Magnesium chloride, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:7786-30-3
:
Formula:MgCl₂
MDL Number:MFCD00011106
:95.21 g/mol
Merck Index:13,05684
Boiling Pt:1412 °C (1013 hPa)
Melting Pt:715 °C
Density:2.32 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:232-094-6