
Sodium metavanadate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:13718-26-8
:
Formula:NaO₃V
MDL Number:MFCD00011125
:121.93 g/mol
Merck Index:12,08849
Melting Pt:630 °C
Density:5.15 g/cm³ (20 °C)
EINECS:237-272-7
ADR:6.1,III
:M-Clarity™ quality level = MQ200
UN:3285
:GHS06
Storage Temperature:Ambient