Sodium sulphate, Sigma-Aldrich®
Catalog # SIAL204447-100G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
About this item
:
Merck Index:12,08829
Boiling Pt:1429 °C (1013 hPa)
Melting Pt:888 °C
Density:2.7 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
:142.04 g/mol
Storage Temperature:Ambient
EINECS:231-820-9
CAS Number:7757-82-6
Formula:Na₂SO₄
MDL Number:MFCD00003504