Order Entry
Export
ContactUsLinkComponent
Potassium sulphate, Sigma-Aldrich®
Potassium sulphate, Sigma-Aldrich®
Catalog # SIALP8541-1KG
Supplier:  Merck
CAS Number:  
Potassium sulphate, Sigma-Aldrich®
Catalog # SIALP8541-1KG
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

About this item

 :
 :174.26 g/mol
Merck Index:13,07759
Boiling Pt:1698 °C (1013 hPa)
Melting Pt:1069 °C
Density:2.66 g/cm³ (20 °C)
 :M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:231-915-5
CAS Number:7778-80-5
Formula:K₂SO₄
MDL Number:MFCD00011388