Potassium sulphate ≥99.99% (metals basis)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7778-80-5
:
Formula:K₂SO₄
MDL Number:MFCD00011388
:174.26 g/mol
Merck Index:14,7674
Boiling Pt:1689 °C
Melting Pt:1069 °C
Density:2.662
Storage Temperature:Ambient
EINECS:231-915-5