Order Entry
Export
Orders LinkContactUsLinkComponent
 

Cesium sulphate

Supplier:   Roth Carl
Cesium sulphate
Supplier:  Roth Carl
Certificates

2 Options Available Below

Product Details & Documents
  • CAS Number:10294-54-9
  • Formula:Cs₂O₄S
  • MDL Number:MFCD00010959
  • Molecular Weight:361.87 g/mol
  • Merck Index:13,02025
  • Boiling Pt:1627 °C (1013 hPa)
  • Melting Pt:1010 °C
  • Density:4.24 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:233-662-6
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 166,00 €
    166,00 €
    1 * 100 g
       166,00 €

  • Glass bottle for solids
    1 kg
    Log in for availability
    Log in for availability
    1 * 1 kg
    1 * 1 kg - 1 120,00 €
    1 120,00 €
    1 * 1 kg
       1 120,00 €