Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Sodium sulphite
Sodium sulphite, Sigma-Aldrich®
Sodium sulphite, Sigma-Aldrich®
:
Merck
Sodium sulphite, Sigma-Aldrich®
:
Merck
:
7757-83-7
:
:
Na₂SO₃
:
MFCD00003503
:
126.04 g/mol
:
13,08757
:
> 500 °C
:
2.63 g/cm³ (20 °C)
:
M-Clarity™ quality level = MQ200
:
Ambient
:
231-821-4
:
01-2119537420-49
SIALS4672-100G_TDS_EN