Sodium sulphite
Supplier: Roth Carl
Certificates
About this item
CAS Number:7757-83-7
:
Formula:Na₂SO₃
MDL Number:MFCD00003503
:126.04 g/mol
Merck Index:13,08757
Melting Pt:> 500 °C
Density:2.63 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-821-4
REACH:01-2119537420-49