Potassium thiocyanate
Supplier: Roth Carl
Jump to:
About this item
Synonyms: Potassium rhodanide
- CAS Number:333-20-0
- Formula:KSCN
- MDL Number:MFCD00011413
- Molecular Weight:97.18 g/mol
- Merck Index:12,07866
- Boiling Pt:500 °C (1013 hPa)
- Melting Pt:175 °C
- Density:1.61 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:206-370-1
2 Options Available Below
Product Details & Documents
Synonyms: Potassium rhodanide
- CAS Number:333-20-0
- Formula:KSCN
- MDL Number:MFCD00011413
- Molecular Weight:97.18 g/mol
- Merck Index:12,07866
- Boiling Pt:500 °C (1013 hPa)
- Melting Pt:175 °C
- Density:1.61 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:206-370-1



