
Potassium thiocyanate 98%, pure
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:333-20-0
:
Formula:KSCN
MDL Number:MFCD00011413
Merck Index:12,07866
Boiling Pt:500 °C (1013 hPa)
Melting Pt:170…179 °C
Density:1.886 g/cm³
:97.18 g/mol
EINECS:206-370-1
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Colorless to white Crystalline powder or crystals
- Titration Argentometric:≥97.5 %