Order Entry
Export
Orders LinkContactUsLinkComponent
Potassium peroxodisulphate

Potassium peroxodisulphate

Glass bottle for solids, 1 kg
Catalog # ROTH6795.1
Oxidizing
Health
Irritant
Supplier:   Roth Carl
Potassium peroxodisulphate
Catalog # ROTH6795.1
Supplier:  Roth Carl
CAS Number:  7727-21-1
Potassium peroxodisulphate

Glass bottle for solids, 1 kg

CAS Number:  7727-21-1
Price
42,50 €
Log in for availability
1 * 1 kg
Product Details & Documents
Synonyms:  di-Potassium peroxodisulphate, Peroxydisulphuric acid dipotassium salt, Dipotassium peroxodisulphate, Potassium persulphate
  • CAS Number:7727-21-1
  • Formula:K₂S₂O₈
  • MDL Number:MFCD00011386
  • Molecular Weight:270.32 g/mol
  • Merck Index:13,07741
  • Melting Pt:100 °C
  • Density:2.477 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:231-781-8
  • UN:1492
  • ADR:5.1,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg

Specifications