Potassium peroxodisulphate
Glass bottle for solids, 1 kg
Catalog # MOLEM35896452
Supplier: Molekula
Jump to:
Product Details & Documents
Synonyms: di-Potassium peroxodisulphate, Peroxydisulphuric acid dipotassium salt, Dipotassium peroxodisulphate, Potassium persulphate
- CAS Number:7727-21-1
- Formula:K₂S₂O₈
- MDL Number:MFCD00011386
- Molecular Weight:270.32 g/mol
- Merck Index:13,07741
- Melting Pt:100 °C
- Density:2.477 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:231-781-8
- UN:1492
- ADR:5.1,III
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:1 kg



