


Potassium tetrachloroplatinate(II), Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:10025-99-7
:
Formula:Cl₄K₂Pt
MDL Number:MFCD00011378
:415.09 g/mol
Merck Index:12,07855
Boiling Pt:35…40 °C (20 mmHg)
Melting Pt:265 °C
Density:3.38 g/cm³ (20 °C)
UN:3288
ADR:6.1,III
:M-Clarity™ quality level = MQ100
EINECS:233-050-9
:GHS06
:GHS05
:GHS08