Potassium hexacyanoferrate(III)
Glass bottle for solids, 1 kg
Catalog # MOLE19316295-1KG
Supplier: Molekula
Jump to:
Product Details & Documents
Synonyms: Potassium ferricyanide(III), Tripotassium hexacyanoferrate, Potassium ferricyanide
- CAS Number:13746-66-2
- Formula:K₃Fe(CN)₆
- MDL Number:MFCD00011392
- Molecular Weight:329.25 g/mol
- Merck Index:13,07712
- Density:1.85 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:237-323-3
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:1 kg



