Order Entry
Export
Orders LinkContactUsLinkComponent
Cesium chloride

Cesium chloride

Supplier:   Roth Carl
Cesium chloride
Supplier:  Roth Carl
Certificates

2 Options Available Below

Product Details & Documents
Synonyms:  Cesium Chloride
  • CAS Number:7647-17-8
  • Formula:CsCl
  • MDL Number:MFCD00010955
  • Molecular Weight:168.36 g/mol
  • Merck Index:13,02021
  • Boiling Pt:1303 °C (1013 hPa)
  • Melting Pt:646 °C
  • Density:3.7808 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:231-600-2
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 138,00 €
    138,00 €
    1 * 100 g
       138,00 €

  • Glass bottle for solids
    1 kg
    Log in for availability
    Log in for availability
    1 * 1 kg
    1 * 1 kg - 965,00 €
    965,00 €
    1 * 1 kg
       965,00 €