Cesium chloride
Supplier: Roth Carl
Jump to:
About this item
Synonyms: Cesium Chloride
- CAS Number:7647-17-8
- Formula:CsCl
- MDL Number:MFCD00010955
- Molecular Weight:168.36 g/mol
- Merck Index:13,02021
- Boiling Pt:1303 °C (1013 hPa)
- Melting Pt:646 °C
- Density:3.7808 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:231-600-2
2 Options Available Below
Product Details & Documents
Synonyms: Cesium Chloride
- CAS Number:7647-17-8
- Formula:CsCl
- MDL Number:MFCD00010955
- Molecular Weight:168.36 g/mol
- Merck Index:13,02021
- Boiling Pt:1303 °C (1013 hPa)
- Melting Pt:646 °C
- Density:3.7808 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:231-600-2
Product Family Options
Product Information
- Pack typePkAvailabilityPrice



