
Sodium tetraphenylborate ACS
:143-66-8
:
:(C₆H₅)₄BNa
:MFCD00011494
:342.22 g/mol
:13,08765
:>300 °C
:1,15 g/cm³ (20 °C)
:Ambient
:205-605-5
:2811
:6.1,III
:GHS06
- Appearance:White to light beige Powder or crystals
- Infrared spectrum:Conforms
- Loss on drying:≤0.5 % (105°C)
- Gravimetry:≥99.5 %
- Clarity of solution:Passes test