Lithium tetrahydroborate, Sigma-Aldrich®
Catalog # SIAL686026-10G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g
- Seq:3236
Specifications
About this item
:
:21.78 g/mol
Merck Index:13,05547
Boiling Pt:66 °C (1013 hPa)
Melting Pt:274…277 °C
Density:0.666 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:241-021-7
UN:1413
ADR:4.3,I
:M-Clarity™ quality level = MQ100
CAS Number:16949-15-8
Formula:H₄BLi
MDL Number:MFCD00011088