Water, BioPerformance Certified, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
Synonyms: Hydrogen oxide
CAS Number:7732-18-5
:
Formula:H₂O
MDL Number:MFCD00011332
Boiling Pt:100
Melting Pt:0
Density:1 g/cm³ (4 °C)
Molecular Weight:18.02 g/mol
Merck Index:13,10098
:M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:231-791-2
Specifications
- Environmentally Preferable: