Order Entry
Export
Orders LinkContactUsLinkComponent
Molybdic acid ACS

Molybdic acid ACS

Irritant
Health
Molybdic acid ACS
Certificates
Molybdic acid ACS

2 Options Available Below

Product Details & Documents
  • CAS Number:7782-91-4
  • Formula:H₂MoO₄
  • MDL Number:MFCD00036287
  • Molecular Weight:161.95 g/mol
  • Merck Index:13,06262
  • Boiling Pt:~ 300 °C (1013 hPa)
  • Melting Pt:300 °C
  • Density:3.1 g/cm³
  • Storage Temperature:Ambient
  • EINECS:231-970-5
Specifications for Product Family

Specification Test Results

  • Appearance:
    White Crystalline powder or crystals
  • Titration:
    ≥85.0 % (Precipitation titraton of molybdate)
  • Chloride (Cl):
    ≤0.002 %
  • Phosphate (PO4):
    ≤5 ppm
  • Sulfate (SO4):
    ≤0.2 %
  • AsO4, PO4, SiO4 (as SiO2):
    ≤0.001 %
  • Heavy metals (as Pb):
    ≤0.003 %
  • Insoluble matter:
    ≤0.01 % (in dilute NH4OH)
Product Family Options

Product Information