1,3-Dithiane, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:505-23-7
:
Formula:C₄H₈S₂
MDL Number:MFCD00006654
:120.24 g/mol
Boiling Pt:207…208 °C (1013 hPa)
Melting Pt:51…54 °C
Density:1.36 g/cm³ (25 °C)
EINECS:208-006-7
ADR:9,
:M-Clarity™ quality level = MQ100
UN:3335