Order Entry
Export
Orders LinkContactUsLinkComponent
1,3-Dioxane, Sigma-Aldrich®

1,3-Dioxane, Sigma-Aldrich®

Glass bottle for solids, 5 g
Catalog # SIAL283061-5G
Supplier:   Merck
1,3-Dioxane, Sigma-Aldrich®
Catalog # SIAL283061-5G
Supplier:  Merck
CAS Number:  505-22-6
1,3-Dioxane, Sigma-Aldrich®

Glass bottle for solids, 5 g

CAS Number:  505-22-6
Price
131,00 €
Log in for availability
1 * 5 g
Product Details & Documents
Synonyms:  m-Dioxane, 1,3-Dioxacyclohexane
  • CAS Number:505-22-6
  • Formula:C₄H₈O₂
  • MDL Number:MFCD00006566
  • Molecular Weight:88.11 g/mol
  • Boiling Pt:105 °C (1013 hPa)
  • Melting Pt:–74.2 °C
  • Density:1.0399 g/cm³ (15 °C)
  • Storage Temperature:Ambient
  • EINECS:208-005-1
  • UN:1165
  • ADR:3,II
  •  :M-Clarity™ quality level = MQ200
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications