Fluorescein disodium salt C.I. 45350, J.T.Baker®
Catalog # 1314.0025
Supplier: Avantor
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance of solution:passes test
Specification Test Results
About this item
:
:376.28 g/mol
Merck Index:13,04185
Melting Pt:> 300 °C
Density:1.601 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:208-253-0
CAS Number:518-47-8
Formula:C₂₀H₁₀Na₂O₅
MDL Number:MFCD00167039