Fluorescein
1 KIT
Catalog # ROCH11684795910
Supplier: ROCHE
Jump to:
Product Details & Documents
Synonyms: 3',6'-Dihydroxy-3H-spiro[isobenzofuran-1,9'-xanthen]-3-one, 2-(6-Hydroxy-3-oxo-(3H)-xanthen-9-yl)benzoic acid
- CAS Number:2321-07-5
- Formula:C₂₀H₁₂O₅
- MDL Number:MFCD00005050
- Molecular Weight:332.31 g/mol
- Merck Index:13,04185
- Melting Pt:314…316 °C
- :M-Clarity™ quality level = MQ100
- Storage Temperature:Ambient
- EINECS:219-031-8
Specifications
Specifications
- Pk:1 KIT



