Order Entry
Export
Orders LinkContactUsLinkComponent
Fluorescein

Fluorescein

Glass bottle for solids, 100 g
Catalog # MOLEM49740067
Supplier:   Molekula
Fluorescein
Catalog # MOLEM49740067
Supplier:  Molekula
CAS Number:  2321-07-5
Fluorescein
Fluorescein

Glass bottle for solids, 100 g

CAS Number:  2321-07-5
Price
Inquire For Price
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  3',6'-Dihydroxy-3H-spiro[isobenzofuran-1,9'-xanthen]-3-one, 2-(6-Hydroxy-3-oxo-(3H)-xanthen-9-yl)benzoic acid
  • CAS Number:2321-07-5
  • Formula:C₂₀H₁₂O₅
  • MDL Number:MFCD00005050
  • Molecular Weight:332.31 g/mol
  • Merck Index:13,04185
  • Melting Pt:314…316 °C
  • Storage Temperature:Ambient
  • EINECS:219-031-8
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications