Fluorescein ≥90%
Supplier: Thermo Fisher Scientific
Jump to:
About this item
Purity: 90+% CAS No.: 2321-07-5 Molecular Formula: C20H12O5 Formula Weight: 332.32 Melting Point: 320° Beilstein Reference: 94324 Merck Reference: 14,4159 MDL No.: MFCD00005050
Synonyms: 3',6'-Dihydroxy-3H-spiro[isobenzofuran-1,9'-xanthen]-3-one, 2-(6-Hydroxy-3-oxo-(3H)-xanthen-9-yl)benzoic acid
- CAS Number:2321-07-5
- Formula:C₂₀H₁₂O₅
- MDL Number:MFCD00005050
- Molecular Weight:332.31 g/mol
- Merck Index:14,4159
- Melting Pt:320 °C
- Storage Temperature:Ambient
- EINECS:219-031-8
2 Options Available Below
Product Details & Documents
Purity: 90+% CAS No.: 2321-07-5 Molecular Formula: C20H12O5 Formula Weight: 332.32 Melting Point: 320° Beilstein Reference: 94324 Merck Reference: 14,4159 MDL No.: MFCD00005050
Synonyms: 3',6'-Dihydroxy-3H-spiro[isobenzofuran-1,9'-xanthen]-3-one, 2-(6-Hydroxy-3-oxo-(3H)-xanthen-9-yl)benzoic acid
- CAS Number:2321-07-5
- Formula:C₂₀H₁₂O₅
- MDL Number:MFCD00005050
- Molecular Weight:332.31 g/mol
- Merck Index:14,4159
- Melting Pt:320 °C
- Storage Temperature:Ambient
- EINECS:219-031-8



